Product Details

SNP ID
rs199946533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:37993919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAACCGGAAGATGGTGCCGGTA[C/T]CCGAGGGGGCTTACGGGAACTTTTT
Phenotype
MIM: 608592
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTDSPL PubMed Links

Gene Details

Gene
CTDSPL
Gene Name
CTD small phosphatase like
There are no transcripts associated with this gene.

Gene
VILL
Gene Name
villin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015873.3 432 Missense Mutation CCC,TCC P28S NP_056957.3
XM_005265191.3 432 Missense Mutation CCC,TCC P28S XP_005265248.1
XM_005265192.4 432 Missense Mutation CCC,TCC P28S XP_005265249.1
XM_006713184.3 432 Missense Mutation CCC,TCC P28S XP_006713247.1
XM_011533770.2 432 Missense Mutation CCC,TCC P28S XP_011532072.1
XM_011533771.1 432 Missense Mutation CCC,TCC P28S XP_011532073.1
XM_011533773.2 432 Missense Mutation CCC,TCC P28S XP_011532075.1

View Full Product Details