Product Details
- SNP ID
-
rs200205387
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:52435607 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACCCCTCCTCTGCCCCCTTCTAC[A/G]TGGCCTCCACCTCCCGGGGCGTCCG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SEMA3G
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61729091] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SEMA3G
- Gene Name
- semaphorin 3G
There are no transcripts associated with this gene.
View Full Product Details