Product Details

SNP ID
rs200017985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98087671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGTTCATGTGAGTTTACTATTG[C/T]GACTAACTTTCTGCAGGTTTAACAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5AC2 PubMed Links

Gene Details

Gene
OR5AC2
Gene Name
olfactory receptor family 5 subfamily AC member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054106.1 499 Nonsense Mutation CGA,TGA R167* NP_473447.1

View Full Product Details