Product Details

SNP ID
rs200481823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49360215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCTTGCAGCAAGGTTTCACAA[A/G]ACAAGGCACCCAGATTTTTTCTTCC
Phenotype
MIM: 138320 MIM: 165390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPX1 PubMed Links

Gene Details

Gene
GPX1
Gene Name
glutathione peroxidase 1
There are no transcripts associated with this gene.

Gene
RHOA
Gene Name
ras homolog family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313941.1 733 Silent Mutation GTC,GTT V192V NP_001300870.1
NM_001313943.1 733 UTR 3 NP_001300872.1
NM_001313944.1 733 Silent Mutation GTC,GTT V172V NP_001300873.1
NM_001313945.1 733 Silent Mutation GTC,GTT V111V NP_001300874.1
NM_001313946.1 733 Silent Mutation GTC,GTT V108V NP_001300875.1
NM_001313947.1 733 UTR 3 NP_001300876.1
NM_001664.3 733 Silent Mutation GTC,GTT V192V NP_001655.1

View Full Product Details