Product Details

SNP ID
rs200260896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169822150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTTTAAAGAACATCAGGGTCCTC[C/T]ACCTGGATAAGAACAACCTGAGGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC34 PubMed Links

Gene Details

Gene
LRRC34
Gene Name
leucine rich repeat containing 34
There are no transcripts associated with this gene.

Gene
LRRIQ4
Gene Name
leucine rich repeats and IQ motif containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080460.1 340 Missense Mutation CAC,TAC H77Y NP_001073929.1
XM_005247421.2 340 Missense Mutation CAC,TAC H77Y XP_005247478.1
XM_006713613.3 340 Missense Mutation CAC,TAC H77Y XP_006713676.1
XM_011512748.1 340 Missense Mutation CAC,TAC H77Y XP_011511050.1
XM_011512749.1 340 Missense Mutation CAC,TAC H77Y XP_011511051.1

View Full Product Details