Product Details

SNP ID
rs199903675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:155833525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAGTAATATATAGGGAATCCCCC[C/T]TGATGTTCTACTTTAGGAGTCCACC
Phenotype
MIM: 603690
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC33A1 PubMed Links

Gene Details

Gene
SLC33A1
Gene Name
solute carrier family 33 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190992.1 1754 Silent Mutation CAA,CAG Q403Q NP_001177921.1
NM_004733.3 1754 Silent Mutation CAA,CAG Q403Q NP_004724.1
XM_011513311.2 1754 Intron XP_011511613.1
XM_017007462.1 1754 Intron XP_016862951.1
XM_017007463.1 1754 Silent Mutation CAA,CAG Q233Q XP_016862952.1
XM_017007464.1 1754 Silent Mutation CAA,CAG Q132Q XP_016862953.1

View Full Product Details