Product Details

SNP ID
rs199936836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:194397411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGTTCAGCCACAGCTCCTGCAGG[C/T]CCCCCATCTCCCCGAAGAGCACCCC
Phenotype
MIM: 610232 MIM: 173511
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP13A3 PubMed Links

Gene Details

Gene
ATP13A3
Gene Name
ATPase 13A3
There are no transcripts associated with this gene.

Gene
GP5
Gene Name
glycoprotein V platelet
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004488.2 903 Missense Mutation NP_004479.1

View Full Product Details