Product Details
- SNP ID
-
rs200706140
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:82928225 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCTGCCTTTGATTTTCCCTTCTT[G/T]TCTGCCTGCTCTGCCTGGGCAAGGA
- Phenotype
-
MIM: 613367
MIM: 612537
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LIN54
PubMed Links
Gene Details
- Gene
- LIN54
- Gene Name
- lin-54 DREAM MuvB core complex component
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001115007.2 |
2052 |
Missense Mutation |
GAA,GAC |
E488D |
NP_001108479.1 |
NM_001115008.2 |
2052 |
Missense Mutation |
GAA,GAC |
E488D |
NP_001108480.1 |
NM_001288996.1 |
2052 |
Missense Mutation |
GAA,GAC |
E620D |
NP_001275925.1 |
NM_001288997.1 |
2052 |
Missense Mutation |
GAA,GAC |
E488D |
NP_001275926.1 |
NM_194282.3 |
2052 |
Missense Mutation |
GAA,GAC |
E709D |
NP_919258.2 |
XM_005262750.4 |
2052 |
Missense Mutation |
GAA,GAC |
E709D |
XP_005262807.1 |
XM_006714081.3 |
2052 |
Missense Mutation |
GAA,GAC |
E709D |
XP_006714144.1 |
XM_017007728.1 |
2052 |
Missense Mutation |
GAA,GAC |
E620D |
XP_016863217.1 |
XM_017007729.1 |
2052 |
Missense Mutation |
GAA,GAC |
E308D |
XP_016863218.1 |
XM_017007730.1 |
2052 |
Missense Mutation |
GAA,GAC |
E308D |
XP_016863219.1 |
- Gene
- THAP9
- Gene Name
- THAP domain containing 9
There are no transcripts associated with this gene.
View Full Product Details