Product Details

SNP ID
rs200098299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:94299644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAGTAGAAGTCTGCCCAAGTTA[C/G]CTAGTTTAAAGGAAACAAACTTTTC
Phenotype
MIM: 602598 MIM: 612761
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HPGDS PubMed Links

Gene Details

Gene
HPGDS
Gene Name
hematopoietic prostaglandin D synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014485.2 527 Missense Mutation CTA,GTA L146V NP_055300.1
XM_005262932.2 527 Missense Mutation CTA,GTA L115V XP_005262989.1
Gene
SMARCAD1
Gene Name
SWI/SNF-related, matrix-associated actin-dependent regulator of chromatin, subfamily a, containing DEAD/H box 1
There are no transcripts associated with this gene.

View Full Product Details