Product Details

SNP ID
rs200627228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:114827829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCTCCAGTGCTATCTCACTTTC[C/T]GCAGTTCCTGTCTCAGCCACGATGG
Phenotype
MIM: 615039
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDST4 PubMed Links

Gene Details

Gene
NDST4
Gene Name
N-deacetylase and N-sulfotransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022569.2 2283 Missense Mutation CAG,CGG Q869R NP_072091.1
XM_017008545.1 2283 Missense Mutation CAG,CGG Q490R XP_016864034.1
XM_017008546.1 2283 Missense Mutation CAG,CGG Q490R XP_016864035.1

View Full Product Details