Product Details

SNP ID
rs200133015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67822389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAGGTAGGCTGTCACTCGAGTA[C/T]ACACTCCTGGCTTATCCGGCAGGCC
Phenotype
MIM: 605369
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS11D PubMed Links

Gene Details

Gene
TMPRSS11D
Gene Name
transmembrane protease, serine 11D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004262.2 1270 Missense Mutation NP_004253.1
XM_005265710.4 1270 Missense Mutation XP_005265767.1
XM_017008851.1 1270 Missense Mutation XP_016864340.1

View Full Product Details