Product Details

SNP ID
rs200495317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:121036855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGTGAATAAAGAAGGTGACTTT[C/T]TGGTGAGAAGAGACTGGAGCAAACC
Phenotype
MIM: 616506
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDNF PubMed Links

Gene Details

Gene
NDNF
Gene Name
neuron derived neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024574.3 1643 Silent Mutation CAA,CAG Q372Q NP_078850.3

View Full Product Details