Product Details

SNP ID
rs200412722
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69957603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTATCGTTGGAGATTTAAGGACAG[A/G]CATCACTCTGCCCTTAGTGTAGACA
Phenotype
MIM: 115460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSN2 PubMed Links

Gene Details

Gene
CSN2
Gene Name
casein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302770.1 362 Missense Mutation CCT,TCT P115S NP_001289699.1
NM_001891.3 362 Missense Mutation CCT,TCT P116S NP_001882.1
XM_017007760.1 362 Missense Mutation CCT,TCT P116S XP_016863249.1

View Full Product Details