Product Details

SNP ID
rs200618308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69998462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAAGTTCCTTGTCTTTGCCTTC[A/T]TCTTGGCTCTCATGGTTTCCATGAT
Phenotype
MIM: 184470
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
STATH PubMed Links

Gene Details

Gene
STATH
Gene Name
statherin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009181.1 135 Missense Mutation ATC,TTC I9F NP_001009181.1
NM_003154.2 135 Missense Mutation ATC,TTC I9F NP_003145.1

View Full Product Details