Product Details
- SNP ID
-
rs200781949
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:73436288 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAAGTGGGTGGAATCAATTTTTT[A/T]AATTTTCCTACTAAATTTTACTGAA
- Phenotype
-
MIM: 104150
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AFP
PubMed Links
Gene Details
- Gene
- AFP
- Gene Name
- alpha fetoprotein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001134.2 |
70 |
Nonsense Mutation |
TAA,TTA |
*9L |
NP_001125.1 |
View Full Product Details