Product Details
- SNP ID
-
rs200958105
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:46734983 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCACTCTTTTGGGGTAACTCTGCC[A/G]ACAGGGGATAGGTTCCATTCTATTC
- Phenotype
-
MIM: 609811
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
COX7B2
PubMed Links
Gene Details
- Gene
- COX7B2
- Gene Name
- cytochrome c oxidase subunit 7B2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_130902.2 |
279 |
Silent Mutation |
GTC,GTT |
V70V |
NP_570972.2 |
XM_005248056.4 |
279 |
Silent Mutation |
GTC,GTT |
V70V |
XP_005248113.1 |
XM_011513630.2 |
279 |
Silent Mutation |
GTC,GTT |
V70V |
XP_011511932.1 |
XM_011513631.2 |
279 |
Silent Mutation |
GTC,GTT |
V70V |
XP_011511933.1 |
XM_011513632.2 |
279 |
Silent Mutation |
GTC,GTT |
V70V |
XP_011511934.1 |
XM_011513633.2 |
279 |
Silent Mutation |
GTC,GTT |
V70V |
XP_011511935.1 |
XM_011513634.2 |
279 |
Silent Mutation |
GTC,GTT |
V70V |
XP_011511936.1 |
XM_011513635.2 |
279 |
Silent Mutation |
GTC,GTT |
V70V |
XP_011511937.1 |
XM_011513636.1 |
279 |
Silent Mutation |
GTC,GTT |
V70V |
XP_011511938.1 |
XM_011513637.2 |
279 |
Silent Mutation |
GTC,GTT |
V70V |
XP_011511939.1 |
XM_011513638.2 |
279 |
Silent Mutation |
GTC,GTT |
V70V |
XP_011511940.1 |
View Full Product Details