Product Details
- SNP ID
-
rs200464150
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:76313223 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTACTTTCTGTATCCTCTTTTCTT[G/T]ATTTCTGATCATTGAAGACATTGCT
- Phenotype
-
MIM: 607406
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC158
PubMed Links
Gene Details
- Gene
- CCDC158
- Gene Name
- coiled-coil domain containing 158
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001042784.1 |
3442 |
Missense Mutation |
AAA,CAA |
K1097Q |
NP_001036249.1 |
XM_005262974.4 |
3442 |
Missense Mutation |
AAA,CAA |
K1097Q |
XP_005263031.1 |
XM_006714204.3 |
3442 |
Missense Mutation |
AAA,CAA |
K1101Q |
XP_006714267.1 |
XM_011531908.2 |
3442 |
Missense Mutation |
AAA,CAA |
K1101Q |
XP_011530210.1 |
XM_011531909.2 |
3442 |
Missense Mutation |
AAA,CAA |
K1101Q |
XP_011530211.1 |
XM_011531910.2 |
3442 |
Missense Mutation |
AAA,CAA |
K1101Q |
XP_011530212.1 |
XM_011531911.1 |
3442 |
Missense Mutation |
AAA,CAA |
K1101Q |
XP_011530213.1 |
XM_011531912.2 |
3442 |
Missense Mutation |
AAA,CAA |
K1101Q |
XP_011530214.1 |
XM_011531913.1 |
3442 |
Missense Mutation |
AAA,CAA |
K1101Q |
XP_011530215.1 |
XM_011531914.2 |
3442 |
Missense Mutation |
AAA,CAA |
K1101Q |
XP_011530216.1 |
XM_011531915.2 |
3442 |
Missense Mutation |
AAA,CAA |
K1050Q |
XP_011530217.1 |
XM_011531916.2 |
3442 |
Missense Mutation |
AAA,CAA |
K1019Q |
XP_011530218.1 |
XM_011531917.1 |
3442 |
Missense Mutation |
AAA,CAA |
K1019Q |
XP_011530219.1 |
XM_017008160.1 |
3442 |
Missense Mutation |
AAA,CAA |
K1101Q |
XP_016863649.1 |
XM_017008161.1 |
3442 |
Missense Mutation |
AAA,CAA |
K801Q |
XP_016863650.1 |
- Gene
- FAM47E-STBD1
- Gene Name
- FAM47E-STBD1 readthrough
There are no transcripts associated with this gene.
- Gene
- STBD1
- Gene Name
- starch binding domain 1
There are no transcripts associated with this gene.
View Full Product Details