Product Details

SNP ID
rs200748598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:140389288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCCATCTCCAGATCCTGTGCTC[C/T]CATCTGCTTCTTTCATCTAGAAAAA
Phenotype
MIM: 601858
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLGN PubMed Links

Gene Details

Gene
CLGN
Gene Name
calmegin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130675.1 1903 Missense Mutation GAG,GGG E590G NP_001124147.1
NM_004362.2 1903 Missense Mutation GAG,GGG E590G NP_004353.1
Gene
SCOC
Gene Name
short coiled-coil protein
There are no transcripts associated with this gene.

View Full Product Details