Product Details

SNP ID
rs200415043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:144646946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAGCCCGGGGCTAGGGCGCCTG[A/G]AGAATAAGGTAGGCACTCACCGGCT
Phenotype
MIM: 606178
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HHIP PubMed Links

Gene Details

Gene
HHIP
Gene Name
hedgehog interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022475.2 820 Missense Mutation AAG,GAG K91E NP_071920.1
XM_005263178.4 820 Missense Mutation AAG,GAG K91E XP_005263235.1
XM_006714288.3 820 Missense Mutation AAG,GAG K91E XP_006714351.1
Gene
HHIP-AS1
Gene Name
HHIP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details