Product Details

SNP ID
rs200882978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154584826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAGGATGTGTTTGGAGGACATC[A/C]TCACAGTCTAAAAAAAAAATTAAGC
Phenotype
MIM: 134820
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FGA PubMed Links

Gene Details

Gene
FGA
Gene Name
fibrinogen alpha chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000508.4 1978 Missense Mutation GAG,GAT E633D NP_000499.1
NM_021871.3 1978 Intron NP_068657.1

View Full Product Details