Product Details

SNP ID
rs200281108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:121036576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAGTGCCTAGCCAAGCCACGGT[C/G]GCTGAGGAACAGGTACGGAGCTTGT
Phenotype
MIM: 616506
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NDNF PubMed Links

Gene Details

Gene
NDNF
Gene Name
neuron derived neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024574.3 1922 Silent Mutation GCC,GCG A465A NP_078850.3

View Full Product Details