Product Details
- SNP ID
-
rs200638200
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:139060534 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACACTCGTTCTAATGATTTTTCA[C/T]CAGTAGTTCCTGCTAAATCTTCATT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ELF2
PubMed Links
Gene Details
- Gene
- ELF2
- Gene Name
- E74 like ETS transcription factor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001276457.1 |
991 |
Missense Mutation |
GAT,GGT |
D227G |
NP_001263386.1 |
NM_001276458.1 |
991 |
Missense Mutation |
GAT,GGT |
D244G |
NP_001263387.1 |
NM_001276459.1 |
991 |
Missense Mutation |
GAT,GGT |
D215G |
NP_001263388.1 |
NM_006874.3 |
991 |
Missense Mutation |
GAT,GGT |
D256G |
NP_006865.1 |
NM_201999.2 |
991 |
Missense Mutation |
GAT,GGT |
D304G |
NP_973728.1 |
XM_005262803.2 |
991 |
Missense Mutation |
GAT,GGT |
D316G |
XP_005262860.1 |
XM_005262804.2 |
991 |
Missense Mutation |
GAT,GGT |
D304G |
XP_005262861.1 |
XM_005262805.2 |
991 |
Missense Mutation |
GAT,GGT |
D275G |
XP_005262862.1 |
XM_006714128.2 |
991 |
Missense Mutation |
GAT,GGT |
D287G |
XP_006714191.1 |
XM_006714129.3 |
991 |
Missense Mutation |
GAT,GGT |
D212G |
XP_006714192.1 |
XM_011531710.2 |
991 |
Missense Mutation |
GAT,GGT |
D212G |
XP_011530012.1 |
XM_011531711.2 |
991 |
Missense Mutation |
GAT,GGT |
D212G |
XP_011530013.1 |
XM_011531712.2 |
991 |
Missense Mutation |
GAT,GGT |
D212G |
XP_011530014.1 |
XM_011531714.2 |
991 |
Missense Mutation |
GAT,GGT |
D58G |
XP_011530016.1 |
XM_017007856.1 |
991 |
Missense Mutation |
GAT,GGT |
D200G |
XP_016863345.1 |
XM_017007857.1 |
991 |
Missense Mutation |
GAT,GGT |
D200G |
XP_016863346.1 |
XM_017007858.1 |
991 |
Missense Mutation |
GAT,GGT |
D183G |
XP_016863347.1 |
XM_017007859.1 |
991 |
Missense Mutation |
GAT,GGT |
D171G |
XP_016863348.1 |
XM_017007860.1 |
991 |
Missense Mutation |
GAT,GGT |
D171G |
XP_016863349.1 |
XM_017007861.1 |
991 |
Missense Mutation |
GAT,GGT |
D171G |
XP_016863350.1 |
View Full Product Details