Product Details

SNP ID
rs200638397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68670105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTAGCCAACAGAGAATCGAAGAC[G/T]GTACAGAAAGGGTATGTTAAATAGT
Phenotype
MIM: 600069
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UGT2B15 PubMed Links

Gene Details

Gene
UGT2B15
Gene Name
UDP glucuronosyltransferase family 2 member B15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076.3 672 Missense Mutation AGT,CGT S172R NP_001067.2

View Full Product Details