Product Details
- SNP ID
-
rs199739916
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:41360907 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGCCCGAGCCGCCCCCCGAGCCC[A/G]CCTTCTCCGAGGCGCAGAAGTGGAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LIMCH1
PubMed Links
Gene Details
- Gene
- LIMCH1
- Gene Name
- LIM and calponin homology domains 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001112717.2 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
NP_001106188.1 |
NM_001112718.2 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
NP_001106189.1 |
NM_001112719.2 |
123 |
Intron |
|
|
NP_001106190.1 |
NM_001112720.2 |
123 |
Intron |
|
|
NP_001106191.1 |
NM_001289122.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
NP_001276051.1 |
NM_001289124.1 |
123 |
Intron |
|
|
NP_001276053.1 |
NM_014988.3 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
NP_055803.2 |
XM_005248057.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_005248114.1 |
XM_005248058.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_005248115.1 |
XM_005248060.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_005248117.1 |
XM_005248061.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_005248118.1 |
XM_005248067.2 |
123 |
Intron |
|
|
XP_005248124.1 |
XM_005248072.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_005248129.1 |
XM_005248074.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_005248131.1 |
XM_005248075.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_005248132.1 |
XM_006713996.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_006714059.1 |
XM_011513642.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_011511944.1 |
XM_011513643.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_011511945.1 |
XM_011513644.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_011511946.1 |
XM_011513645.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_011511947.1 |
XM_011513646.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_011511948.1 |
XM_011513648.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_011511950.1 |
XM_011513649.1 |
123 |
Intron |
|
|
XP_011511951.1 |
XM_011513651.2 |
123 |
Intron |
|
|
XP_011511953.1 |
XM_011513653.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_011511955.1 |
XM_011513654.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_011511956.1 |
XM_011513655.1 |
123 |
Intron |
|
|
XP_011511957.1 |
XM_011513656.1 |
123 |
Intron |
|
|
XP_011511958.1 |
XM_011513657.1 |
123 |
Intron |
|
|
XP_011511959.1 |
XM_011513658.2 |
123 |
Intron |
|
|
XP_011511960.1 |
XM_017007894.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_016863383.1 |
XM_017007895.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_016863384.1 |
XM_017007896.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_016863385.1 |
XM_017007897.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_016863386.1 |
XM_017007898.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_016863387.1 |
XM_017007899.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_016863388.1 |
XM_017007900.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_016863389.1 |
XM_017007901.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_016863390.1 |
XM_017007902.1 |
123 |
Missense Mutation |
ACC,GCC |
T23A |
XP_016863391.1 |
XM_017007903.1 |
123 |
Intron |
|
|
XP_016863392.1 |
XM_017007904.1 |
123 |
Intron |
|
|
XP_016863393.1 |
XM_017007905.1 |
123 |
Intron |
|
|
XP_016863394.1 |
View Full Product Details