Product Details

SNP ID
rs199879494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:143577652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCACTCTGCTTACAGTCCGTGA[C/T]GGTGTCCTCGATGAAAATGGTAGTT
Phenotype
MIM: 608946
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FREM3 PubMed Links

Gene Details

Gene
FREM3
Gene Name
FRAS1 related extracellular matrix 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168235.1 6379 Missense Mutation ATC,GTC I2127V NP_001161707.1

View Full Product Details