Product Details

SNP ID
rs199577367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99549214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAATCACTGTCCGATCCACCTTC[C/T]TCCATCCTGACAGACTGATTGTCAT
Phenotype
MIM: 616013
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C4orf17 PubMed Links

Gene Details

Gene
C4orf17
Gene Name
chromosome 4 open reading frame 17
There are no transcripts associated with this gene.

Gene
TRMT10A
Gene Name
tRNA methyltransferase 10A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134665.2 1041 Silent Mutation GAA,GAG E298E NP_001128137.1
NM_001134666.2 1041 Silent Mutation GAA,GAG E298E NP_001128138.1
NM_152292.4 1041 Silent Mutation GAA,GAG E298E NP_689505.1
XM_005263352.3 1041 Silent Mutation GAA,GAG E298E XP_005263409.1
XM_006714417.2 1041 Silent Mutation GAA,GAG E298E XP_006714480.1

View Full Product Details