Product Details

SNP ID
rs199803124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:51995075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAACCTCGCTGTAGTGGGCTGA[A/G]AGCGCTTCCTCAGCCCTTGCCCCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC66 PubMed Links

Gene Details

Gene
LRRC66
Gene Name
leucine rich repeat containing 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024611.2 3681 Silent Mutation CTC,CTT L649L NP_001019782.1
XM_011534383.2 3681 Silent Mutation CTC,CTT L701L XP_011532685.1
XM_017008162.1 3681 Silent Mutation CTC,CTT L413L XP_016863651.1

View Full Product Details