Product Details

SNP ID
rs201496236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:188002997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATGAGCCTGGCCCTCAGGCTCTC[A/G]GAGGGGATGATTTCTCAGACTGTTA
Phenotype
MIM: 614572
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFP42 PubMed Links

Gene Details

Gene
ZFP42
Gene Name
ZFP42 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304358.1 432 Missense Mutation AGA,GGA R64G NP_001291287.1
NM_174900.4 432 Intron NP_777560.2
XM_011531604.2 432 Missense Mutation AGA,GGA R64G XP_011529906.1
XM_011531605.1 432 Missense Mutation AGA,GGA R64G XP_011529907.1
XM_011531606.1 432 Missense Mutation AGA,GGA R64G XP_011529908.1
XM_011531607.2 432 Missense Mutation AGA,GGA R64G XP_011529909.1

View Full Product Details