Product Details

SNP ID
rs201841340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:143186006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCAGTGCGTGTCCACCCAGGAA[A/G]GAGAGCTGCGGGAATATGTCTCCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105377623 PubMed Links

Gene Details

Gene
LOC105377623
Gene Name
uncharacterized LOC105377623
There are no transcripts associated with this gene.

Gene
USP38
Gene Name
ubiquitin specific peptidase 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290325.1 1090 Missense Mutation AGA,GGA R186G NP_001277254.1
NM_001290326.1 1090 UTR 5 NP_001277255.1
NM_032557.6 1090 Missense Mutation AGA,GGA R186G NP_115946.2
XM_011532360.2 1090 Missense Mutation AGA,GGA R186G XP_011530662.1

View Full Product Details