Product Details

SNP ID
rs202020493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:83456159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAACGGCTGCTGTCAGCCTTACA[C/T]ATCCCGGGACTCACACGGGTAAAGT
Phenotype
MIM: 611143 MIM: 606769 MIM: 611983
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM175A PubMed Links

Gene Details

Gene
FAM175A
Gene Name
family with sequence similarity 175 member A
There are no transcripts associated with this gene.

Gene
HELQ
Gene Name
helicase, POLQ-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297755.1 228 Intron NP_001284684.1
NM_001297756.1 228 Intron NP_001284685.1
NM_001297757.1 228 Intron NP_001284686.1
NM_001297758.1 228 Intron NP_001284687.1
NM_001297759.1 228 Intron NP_001284688.1
NM_133636.3 228 Intron NP_598375.2
XM_005262711.1 228 Intron XP_005262768.1
XM_005262713.2 228 Intron XP_005262770.1
XM_006714076.2 228 Intron XP_006714139.1
XM_011531580.2 228 Intron XP_011529882.1
XM_017007679.1 228 Intron XP_016863168.1
XM_017007680.1 228 Intron XP_016863169.1
XM_017007681.1 228 Intron XP_016863170.1
XM_017007682.1 228 Intron XP_016863171.1
XM_017007683.1 228 Intron XP_016863172.1
Gene
MRPS18C
Gene Name
mitochondrial ribosomal protein S18C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297767.1 228 Missense Mutation CAT,TAT H28Y NP_001284696.1
NM_001297769.1 228 Missense Mutation CAT,TAT H28Y NP_001284698.1
NM_001297770.1 228 Missense Mutation CAT,TAT H28Y NP_001284699.1
NM_016067.3 228 Missense Mutation CAT,TAT H28Y NP_057151.1

View Full Product Details