Product Details

SNP ID
rs202072717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48074008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGGAGGGGACCACTTGATTGG[A/G]AACTTGGCTCCAAAAGAACTGACAT
Phenotype
MIM: 600058
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TXK PubMed Links

Gene Details

Gene
TXK
Gene Name
TXK tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003328.2 1370 Silent Mutation TTC,TTT F428F NP_003319.2
XM_011513747.2 1370 Silent Mutation TTC,TTT F409F XP_011512049.1
XM_011513748.2 1370 Silent Mutation TTC,TTT F190F XP_011512050.1
XM_017008580.1 1370 Silent Mutation TTC,TTT F431F XP_016864069.1
XM_017008581.1 1370 Silent Mutation TTC,TTT F403F XP_016864070.1
XM_017008582.1 1370 Silent Mutation TTC,TTT F239F XP_016864071.1

View Full Product Details