Product Details

SNP ID
rs201541253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:124669111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATCCCTCCAACACTTCCTGATGC[C/T]CTCTATCTTGCATTGTTCGTGGGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD50 PubMed Links

Gene Details

Gene
ANKRD50
Gene Name
ankyrin repeat domain 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167882.1 5937 Missense Mutation GAG,GGG E1210G NP_001161354.1
NM_020337.2 5937 Missense Mutation GAG,GGG E1389G NP_065070.1
XM_017008471.1 5937 Missense Mutation GAG,GGG E1389G XP_016863960.1

View Full Product Details