Product Details

SNP ID
rs201269910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:127984399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGATGCCAGAATCTGGTTTCAAG[C/T]GATTATCCAGTACACATTGATAAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD18 PubMed Links

Gene Details

Gene
ABHD18
Gene Name
abhydrolase domain containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319305.1 516 UTR 5 NP_001306234.1
NM_001319306.1 516 UTR 5 NP_001306235.1
NM_001319307.1 516 UTR 5 NP_001306236.1
XM_005263238.4 516 Missense Mutation AGC,AGT S51S XP_005263295.1
XM_006714318.3 516 Missense Mutation AGC,AGT S51S XP_006714381.1
XM_006714320.3 516 Missense Mutation AGC,AGT S51S XP_006714383.1
XM_006714322.3 516 Intron XP_006714385.1
XM_011532272.2 516 Missense Mutation AGC,AGT S51S XP_011530574.1
XM_011532273.2 516 Missense Mutation AGC,AGT S51S XP_011530575.1
XM_011532274.2 516 Missense Mutation AGC,AGT S51S XP_011530576.1
XM_011532276.1 516 Missense Mutation AGC,AGT S51S XP_011530578.1
XM_011532278.2 516 UTR 5 XP_011530580.1
XM_011532279.2 516 Intron XP_011530581.1
XM_017008631.1 516 Silent Mutation AGC,AGT S51S XP_016864120.1
XM_017008632.1 516 Intron XP_016864121.1

View Full Product Details