Product Details

SNP ID
rs201390327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:110476438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGAACTTTGCGGAGAGAGAGGG[C/T]TCTAAGAGATACTGCATTCAAACGA
Phenotype
MIM: 138297
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENPEP PubMed Links

Gene Details

Gene
ENPEP
Gene Name
glutamyl aminopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001977.3 366 Silent Mutation GGC,GGT G8G NP_001968.3
XM_017007877.1 366 Intron XP_016863366.1

View Full Product Details