Product Details

SNP ID
rs201801358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1983883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTCTTCTTAGGCTGCTGCTGAG[C/G]AGGGGCCTGGGTCTGCGGGGCCACC
Phenotype
MIM: 606026 MIM: 602952
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR943 PubMed Links

Gene Details

Gene
MIR943
Gene Name
microRNA 943
There are no transcripts associated with this gene.

Gene
NELFA
Gene Name
negative elongation factor complex member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005663.4 1543 Missense Mutation CCT,GCT P434A NP_005654.3
XM_017008589.1 1543 Missense Mutation CCT,GCT P462A XP_016864078.1
Gene
SCARNA22
Gene Name
small Cajal body-specific RNA 22
There are no transcripts associated with this gene.

Gene
WHSC1
Gene Name
Wolf-Hirschhorn syndrome candidate 1
There are no transcripts associated with this gene.

View Full Product Details