Product Details

SNP ID
rs201546209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:8581259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGACACCTTCCTGGCGTCCAAC[G/T]CGGCGCTGAGCGTGGCGGCGCTGAG
Phenotype
MIM: 606921
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR78 PubMed Links

Gene Details

Gene
GPR78
Gene Name
G protein-coupled receptor 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080819.4 770 Missense Mutation GCG,TCG A93S NP_543009.2

View Full Product Details