Product Details

SNP ID
rs201547149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:125317398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACGGTGCAGGCGATGGACAGAGG[C/G]GTGCCTTCCCTCACTGGGCGCGCCG
Phenotype
MIM: 612411
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAT4 PubMed Links

Gene Details

Gene
FAT4
Gene Name
FAT atypical cadherin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291285.1 987 Silent Mutation GGC,GGG G329G NP_001278214.1
NM_001291303.1 987 Silent Mutation GGC,GGG G329G NP_001278232.1
NM_024582.4 987 Silent Mutation GGC,GGG G329G NP_078858.4
XM_011532236.2 987 Silent Mutation GGC,GGG G329G XP_011530538.1
XM_011532237.2 987 Intron XP_011530539.1

View Full Product Details