Product Details

SNP ID
rs201564349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:127883500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTATTAGAAATGTGCCATAATG[C/G]AGAAATGAACAGGTATCTAAAGAAT
Phenotype
MIM: 605031
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PLK4 PubMed Links

Gene Details

Gene
PLK4
Gene Name
polo like kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190799.1 718 Missense Mutation GCA,GGA A63G NP_001177728.1
NM_001190801.1 718 Missense Mutation GCA,GGA A54G NP_001177730.1
NM_014264.4 718 Missense Mutation GCA,GGA A95G NP_055079.3
XM_005262701.2 718 Missense Mutation GCA,GGA A95G XP_005262758.1
XM_017007662.1 718 Missense Mutation GCA,GGA A96G XP_016863151.1
XM_017007663.1 718 Missense Mutation GCA,GGA A96G XP_016863152.1

View Full Product Details