Product Details
- SNP ID
-
rs202164501
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:37960676 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGTCAGTAAAGACCAATGGACCCA[A/G]AAAACAAAAACAGCCAAGCTTTTCT
- Phenotype
-
MIM: 604231
MIM: 609850
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PTTG2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6811863] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PTTG2
- Gene Name
- pituitary tumor-transforming 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006607.2 |
|
Intron |
|
|
NP_006598.2 |
- Gene
- TBC1D1
- Gene Name
- TBC1 domain family member 1
View Full Product Details