Product Details

SNP ID
rs202171658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:103589935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGTCTTGAGCTCTAGCTCATCA[C/T]AGCTGGAAACTTTGATGAAAGGACA
Phenotype
MIM: 162332
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TACR3 PubMed Links

Gene Details

Gene
TACR3
Gene Name
tachykinin receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001059.2 1286 Missense Mutation TAT,TGT Y382C NP_001050.1

View Full Product Details