Product Details

SNP ID
rs202004340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138192505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCACCACCTATAGATAATCACC[G/T]ATAAGCCTGGATAGCAGCAGACGTG
Phenotype
MIM: 603462 MIM: 605664
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDC23 PubMed Links

Gene Details

Gene
CDC23
Gene Name
cell division cycle 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004661.3 1196 Silent Mutation AGA,CGA R389R NP_004652.2
Gene
KIF20A
Gene Name
kinesin family member 20A
There are no transcripts associated with this gene.

View Full Product Details