Product Details
- SNP ID
-
rs202023911
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:138465785 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATGGCCGCGGCCAAGGCCGAGAT[A/G]CAGCTGATGTCCCCGCTGCAGATCT
- Phenotype
-
MIM: 128990
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EGR1
PubMed Links
Gene Details
- Gene
- EGR1
- Gene Name
- early growth response 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001964.2 |
294 |
Missense Mutation |
ATA,ATG |
I8M |
NP_001955.1 |
View Full Product Details