Product Details

SNP ID
rs202163986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150194803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGATGTGGACTTTGCTGCACAC[C/T]GGGGGAACTGGACAGGCAAGCTGGA
Phenotype
MIM: 606205
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC6A7 PubMed Links

Gene Details

Gene
SLC6A7
Gene Name
solute carrier family 6 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014228.4 372 Silent Mutation CGG,TGG R37W NP_055043.2
XM_017009767.1 372 Silent Mutation CGG,TGG R118W XP_016865256.1
XM_017009768.1 372 UTR 5 XP_016865257.1
XM_017009769.1 372 UTR 5 XP_016865258.1
XM_017009770.1 372 UTR 5 XP_016865259.1

View Full Product Details