Product Details

SNP ID
rs202202440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176347748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGCCATTCGGTTGGCCTGGGCA[C/G]GTATCAGTTCCAATGGAGAGGGCTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1191 PubMed Links

Gene Details

Gene
KIAA1191
Gene Name
KIAA1191
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079684.2 Intron NP_001073152.1
NM_001079685.2 Intron NP_001073153.1
NM_001287335.1 Intron NP_001274264.1
NM_001287336.1 Intron NP_001274265.1
NM_020444.4 Intron NP_065177.2
XM_005265941.1 Intron XP_005265998.1
XM_005265945.1 Intron XP_005266002.1
XM_005265946.4 Intron XP_005266003.1
XM_011534595.1 Intron XP_011532897.1
XM_011534596.1 Intron XP_011532898.1
XM_017009651.1 Intron XP_016865140.1
XM_017009652.1 Intron XP_016865141.1
XM_017009653.1 Intron XP_016865142.1
Gene
SIMC1
Gene Name
SUMO interacting motifs containing 1
There are no transcripts associated with this gene.

View Full Product Details