Product Details

SNP ID
rs201791803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69228361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAAATACAGAAGGAAACTTGTG[C/T]CTCAAGAAGAAATGGAATTTATCCA
Phenotype
MIM: 601955 MIM: 611996
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK7 PubMed Links

Gene Details

Gene
CDK7
Gene Name
cyclin dependent kinase 7
There are no transcripts associated with this gene.

Gene
MRPS36
Gene Name
mitochondrial ribosomal protein S36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033281.5 352 Missense Mutation CCT,TCT P90S NP_150597.1

View Full Product Details