Product Details

SNP ID
rs201869203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:56102527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGTAGGGGAGCTAGGTTGTTCC[A/G]AGTGAAAGGAAGATCCCTGATTTTG
Phenotype
MIM: 615189
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD55 PubMed Links

Gene Details

Gene
ANKRD55
Gene Name
ankyrin repeat domain 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024669.2 1156 Missense Mutation CGG,TGG R564W NP_078945.2
XM_011543646.1 1156 Missense Mutation CGG,TGG R337W XP_011541948.1
XM_017009852.1 1156 Missense Mutation CGG,TGG R573W XP_016865341.1
XM_017009853.1 1156 Missense Mutation CGG,TGG R564W XP_016865342.1
XM_017009854.1 1156 Missense Mutation CGG,TGG R411W XP_016865343.1

View Full Product Details