Product Details

SNP ID
rs201914951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1878211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATGGGGTCGTGGAAGACCCCGT[C/T]CACCCAGTTTCTGAGACTGGTTACC
Phenotype
MIM: 606199
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTD-2194D22.4 PubMed Links

Gene Details

Gene
CTD-2194D22.4
Gene Name
uncharacterized LOC101929081
There are no transcripts associated with this gene.

Gene
IRX4
Gene Name
iroquois homeobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278632.1 1536 Missense Mutation AAC,GAC N440D NP_001265561.1
NM_001278633.1 1536 Missense Mutation AAC,GAC N466D NP_001265562.1
NM_001278634.1 1536 Missense Mutation AAC,GAC N440D NP_001265563.1
NM_001278635.1 1536 Missense Mutation AAC,GAC N466D NP_001265564.1
NM_016358.2 1536 Missense Mutation AAC,GAC N440D NP_057442.1

View Full Product Details