Product Details
- SNP ID
-
rs201761413
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:23509998 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCAGGTGGATGACACAGAAGATT[A/C]TGATGAAGAATGGACCCCTAGGCAG
- Phenotype
-
MIM: 609760
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PRDM9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2973615] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRDM9
- Gene Name
- PR domain 9
View Full Product Details