Product Details

SNP ID
rs201998729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176392878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGGTTGAAGTCACATAGCTGGG[C/T]CACCTTCTCCCACTCTGTGCCTGGG
Phenotype
MIM: 118970 MIM: 612861
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL10 PubMed Links

Gene Details

Gene
ARL10
Gene Name
ADP ribosylation factor like GTPase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317948.1 737 Intron NP_001304877.1
NM_173664.5 737 Intron NP_775935.1
XM_011534529.2 737 Intron XP_011532831.1
XM_011534530.2 737 Intron XP_011532832.1
XM_011534531.2 737 Intron XP_011532833.1
XM_017009372.1 737 Intron XP_016864861.1
Gene
CLTB
Gene Name
clathrin light chain B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001834.3 737 Missense Mutation ACC,GCC T178A NP_001825.1
NM_007097.3 737 Missense Mutation ACC,GCC T196A NP_009028.1
XM_017009026.1 737 Missense Mutation ACC,GCC T111A XP_016864515.1
XM_017009027.1 737 Intron XP_016864516.1
Gene
HIGD2A
Gene Name
HIG1 hypoxia inducible domain family member 2A
There are no transcripts associated with this gene.

Gene
NOP16
Gene Name
NOP16 nucleolar protein
There are no transcripts associated with this gene.

View Full Product Details