Product Details

SNP ID
rs202136839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149374460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGGCGGTGGCGGGCAGAGGCGG[C/T]GGCGCACAGGAACCTGGCTGAACAC
Phenotype
MIM: 604627
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL17B PubMed Links

Gene Details

Gene
IL17B
Gene Name
interleukin 17B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317987.1 825 Missense Mutation CAC,CGC H99R NP_001304916.1
NM_014443.2 825 Missense Mutation CAC,CGC H151R NP_055258.1
XM_011537622.2 825 Missense Mutation CAC,CGC H99R XP_011535924.1
XM_017009346.1 825 Missense Mutation CAC,CGC H216R XP_016864835.1
XM_017009347.1 825 Missense Mutation CAC,CGC H99R XP_016864836.1
XM_017009348.1 825 Missense Mutation CAC,CGC H99R XP_016864837.1
Gene
PCYOX1L
Gene Name
prenylcysteine oxidase 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301054.1 825 Intron NP_001287983.1
NM_001301057.1 825 Intron NP_001287986.1
NM_024028.3 825 Intron NP_076933.3
XM_011537680.1 825 Intron XP_011535982.1
XM_011537681.1 825 Intron XP_011535983.1
XM_011537682.2 825 Intron XP_011535984.1

View Full Product Details